About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: NGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77812-917
- 77812-916
- Antigen Symbol:
- NGFR
- NGFR
- Antigen Name:
- Synthetic peptide of NGFR
- Synthetic peptide of NGFR
- Antigen Synonyms:
- CD271|CD271 antigen|Gp80 LNGFR|Gp80-LNGFR|Low affinity nerve growth factor receptor|Low affinity neurotrophin receptor p75NTR|Low-affinity nerve growth factor receptor|Nerve growth factor receptor|Nerve growth factor receptor TNFR superfamily member 16|NGF receptor|Ngfr|p75 ICD|p75 Neurotrophin receptor|p75 NTR|p75(NTR)|p75NTR|TNFR Superfamily Member 16|TNFRSF16|TNR16_HUMAN|Tumor necrosis factor receptor superfamily member 16
- CD271|CD271 antigen|Gp80 LNGFR|Gp80-LNGFR|Low affinity nerve growth factor receptor|Low affinity neurotrophin receptor p75NTR|Low-affinity nerve growth factor receptor|Nerve growth factor receptor|Nerve growth factor receptor TNFR superfamily member 16|NGF receptor|Ngfr|p75 ICD|p75 Neurotrophin receptor|p75 NTR|p75(NTR)|p75NTR|TNFR Superfamily Member 16|TNFRSF16|TNR16_HUMAN|Tumor necrosis factor receptor superfamily member 16
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08138
- P08138
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice