Anti-DHCR24 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: DHCR24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77826-223
- 77826-224
- Antigen Symbol:
- DHCR24
- DHCR24
- Antigen Name:
- Fusion protein of DHCR24
- Fusion protein of DHCR24
- Antigen Synonyms:
- 24 dehydrocholesterol reductase | 24-dehydrocholesterol reductase| 3 beta hydroxysterol delta 24 reductase| 3-beta-hydroxysterol delta-24-reductase| DCE| Delta(24)-sterol reductase| Desmosterol to cholesterol enzyme| DHC24_HUMAN| Dhcr24| Diminuto dwarf1 homolog | Diminuto/dwarf1 homolog| KIAA0018| Nbla03646| Seladin-1| SELADIN1| Selective AD indicator 1| Selective Alzheimer's Disease Indicator 1
- 24 dehydrocholesterol reductase | 24-dehydrocholesterol reductase| 3 beta hydroxysterol delta 24 reductase| 3-beta-hydroxysterol delta-24-reductase| DCE| Delta(24)-sterol reductase| Desmosterol to cholesterol enzyme| DHC24_HUMAN| Dhcr24| Diminuto dwarf1 homolog | Diminuto/dwarf1 homolog| KIAA0018| Nbla03646| Seladin-1| SELADIN1| Selective AD indicator 1| Selective Alzheimer's Disease Indicator 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15392
- Q15392
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together