Anti-PTPN6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: PTPN6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77796-126
- 77796-127
- Antigen Symbol:
- PTPN6
- PTPN6
- Antigen Name:
- Tyrosine-protein phosphatase non-receptor type 6 protein (1-597AA)
- Tyrosine-protein phosphatase non-receptor type 6 protein (1-597AA)
- Antigen Synonyms:
- 70Z-SHP|EC 3.1.3.48|HCP|HCPH|Hematopoietic cell phosphatase|Hematopoietic cell protein tyrosine phosphatase|Hematopoietic cell protein-tyrosine phosphatase|HPTP1C|Protein tyrosine phosphatase 1C|Protein tyrosine phosphatase non receptor type 6|Protein tyrosine phosphatase SHP1|Protein-tyrosine phosphatase 1C|protein-tyrosine phosphatase SHP 1|Protein-tyrosine phosphatase SHP-1|PTN6_HUMAN|PTP 1C|PTP-1C|PTP1C|Ptpn6|SH PTP 1|SH PTP1|SH-PTP1|SHP 1|SHP 1L|SHP1|SHP1L|tyrosine protein phosphatase non receptor type 6|Tyrosine-protein phosphatase non-receptor type 6
- 70Z-SHP|EC 3.1.3.48|HCP|HCPH|Hematopoietic cell phosphatase|Hematopoietic cell protein tyrosine phosphatase|Hematopoietic cell protein-tyrosine phosphatase|HPTP1C|Protein tyrosine phosphatase 1C|Protein tyrosine phosphatase non receptor type 6|Protein tyrosine phosphatase SHP1|Protein-tyrosine phosphatase 1C|protein-tyrosine phosphatase SHP 1|Protein-tyrosine phosphatase SHP-1|PTN6_HUMAN|PTP 1C|PTP-1C|PTP1C|Ptpn6|SH PTP 1|SH PTP1|SH-PTP1|SHP 1|SHP 1L|SHP1|SHP1L|tyrosine protein phosphatase non receptor type 6|Tyrosine-protein phosphatase non-receptor type 6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P29350
- P29350
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together