Anti-DLG1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: DLG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77822-352
- 77822-351
- Antigen Symbol:
- DLG1
- DLG1
- Antigen Name:
- Disks large homolog 1 protein (662-773AA)
- Disks large homolog 1 protein (662-773AA)
- Antigen Synonyms:
- Discs large homolog 1|discs large|Drosophila|homolog of|1|discs|large homolog 1 (Drosophila)|Disks large homolog 1|dJ1061C18.1.1|DKFZp761P0818|DKFZp781B0426|DLG 1|DLG1|DLG1_HUMAN|DLGH 1|DLGH1|hDlg|Presynaptic protein SAP97|SAP 97|SAP-97|SAP97|Synapse associated protein 97|Synapse-associated protein 97
- Discs large homolog 1|discs large|Drosophila|homolog of|1|discs|large homolog 1 (Drosophila)|Disks large homolog 1|dJ1061C18.1.1|DKFZp761P0818|DKFZp781B0426|DLG 1|DLG1|DLG1_HUMAN|DLGH 1|DLGH1|hDlg|Presynaptic protein SAP97|SAP 97|SAP-97|SAP97|Synapse associated protein 97|Synapse-associated protein 97
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12959
- Q12959
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together