Anti-DESI2 Rabbit Polyclonal Antibody
Catalog # 77795-157
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:PPPDE1
- Antigen Symbol:DESI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BSY9
- Antigen Synonyms:DESI2|C1orf121|FAM152A|PPPDE1|CGI-146|PNAS-4|Deubiquitinase DESI2|EC 3.4.19.12|Desumoylating isopeptidase 2|DeSI-2|PPPDE peptidase domain-containing protein 1|Protein FAM152A
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DESI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat