Anti-NIPBL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NIPBL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-275
- 77830-274
- Antigen Symbol:
- NIPBL
- NIPBL
- Antigen Name:
- Nipped-B-like protein (2523-2697AA)
- Nipped-B-like protein (2523-2697AA)
- Antigen Synonyms:
- CDLS|Colon tumor susceptibility 2|Delangin|DKFZp434L1319|FLJ11203|FLJ12597|FLJ13354|FLJ13648|FLJ44854|IDN 3|IDN 3 protein|IDN 3 protein isoform A|IDN 3 protein isoform B|IDN 3B|IDN3 B|IDN3 protein|IDN3 protein isoform A|IDN3 protein isoform B|IDN3B|Mis 4|Mis4|Nipbl|_HUMAN|Nipped B homolog (Drosophila)|Nipped B homolog|Nipped B like|Nipped B like protein|Nipped-B-like protein|Scc 2|SCC 2 homolog|Scc2|SCC2 homolog|Sister chromatid cohesion protein Mis4
- CDLS|Colon tumor susceptibility 2|Delangin|DKFZp434L1319|FLJ11203|FLJ12597|FLJ13354|FLJ13648|FLJ44854|IDN 3|IDN 3 protein|IDN 3 protein isoform A|IDN 3 protein isoform B|IDN 3B|IDN3 B|IDN3 protein|IDN3 protein isoform A|IDN3 protein isoform B|IDN3B|Mis 4|Mis4|Nipbl|_HUMAN|Nipped B homolog (Drosophila)|Nipped B homolog|Nipped B like|Nipped B like protein|Nipped-B-like protein|Scc 2|SCC 2 homolog|Scc2|SCC2 homolog|Sister chromatid cohesion protein Mis4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6KC79
- Q6KC79
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together