Anti-PSMC6 Rabbit Polyclonal Antibody
Catalog # 77795-842
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:SUG2
- Antigen Symbol:PSMC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P62333
- Antigen Synonyms:26S protease regulatory subunit 10B|26S protease regulatory subunit S10B|26S proteasome AAA-ATPase subunit RPT4|CADP44|P44|Proteasome 26S subunit ATPase 6|Proteasome subunit p42|PRS10_HUMAN|PSMC6|Rpt4|SUG2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PSMC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat