Anti-NPHP3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NPHP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-832
- 77831-833
- Antigen Symbol:
- NPHP3
- NPHP3
- Antigen Name:
- Nephrocystin-3 protein (1-130AA)
- Nephrocystin-3 protein (1-130AA)
- Antigen Synonyms:
- Meckel syndrome, type 7|MKS7|Nephrocystin-3|nephronophthisis 3 (adolescent)|NPH3|Nphp3|NPHP3_HUMAN|pcy|RHPD
- Meckel syndrome, type 7|MKS7|Nephrocystin-3|nephronophthisis 3 (adolescent)|NPH3|Nphp3|NPHP3_HUMAN|pcy|RHPD
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z494
- Q7Z494
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together