Anti-CHMP5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:100 to 1:500
Type:
Antigen: CHMP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-050
- 77840-049
- Antigen Symbol:
- CHMP5
- CHMP5
- Antigen Name:
- Charged multivesicular body protein 5 protein (1-219AA)
- Charged multivesicular body protein 5 protein (1-219AA)
- Antigen Synonyms:
- apoptosis-related protein PNAS-2|C9orf83|CGI 34|Charged multivesicular body protein 5|CHMP 5|CHMP family|member 5|chmp5|CHMP5_HUMAN|Chromatin modifying protein 5|Chromatin-modifying protein 5|Chromosome 9 open reading frame 83|HGNC:26942|HSPC177|hVps60|PNAS 2|SNF7 domain containing protein 2|SNF7 domain-containing protein 2|SNF7DC2|Vacuolar protein sorting 60|Vacuolar protein sorting-associated protein 60|Vps60
- apoptosis-related protein PNAS-2|C9orf83|CGI 34|Charged multivesicular body protein 5|CHMP 5|CHMP family|member 5|chmp5|CHMP5_HUMAN|Chromatin modifying protein 5|Chromatin-modifying protein 5|Chromosome 9 open reading frame 83|HGNC:26942|HSPC177|hVps60|PNAS 2|SNF7 domain containing protein 2|SNF7 domain-containing protein 2|SNF7DC2|Vacuolar protein sorting 60|Vacuolar protein sorting-associated protein 60|Vps60
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZZ3
- Q9NZZ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together