Anti-HES5 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: HES5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77807-567
- 77807-568
- Antigen Symbol:
- HES5
- HES5
- Antigen Name:
- Synthetic peptide of HES5
- Synthetic peptide of HES5
- Antigen Synonyms:
- bHLHb38| Class B basic helix-loop-helix protein 38| Hairy and enhancer of split 5| Hairy/Enhancer of spli, Drosophila, homolog of, 5| HES 5| hes family bHLH transcription factor 5| Hes5| HES5 protein| HES5_HUMAN| MGI:104876| Transcription factor HES-5
- bHLHb38| Class B basic helix-loop-helix protein 38| Hairy and enhancer of split 5| Hairy/Enhancer of spli, Drosophila, homolog of, 5| HES 5| hes family bHLH transcription factor 5| Hes5| HES5 protein| HES5_HUMAN| MGI:104876| Transcription factor HES-5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5TA89
- Q5TA89
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together