Anti-CHRAC1 Rabbit Polyclonal Antibody
Catalog # 77806-232
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the C-terminal region of CHRAC15.
- Antigen Symbol:CHRAC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NRG0
- Antigen Synonyms:CHARC1|CHARC15|CHRAC-1|CHRAC-15|Chrac1|CHRAC15|CHRC1_HUMAN|Chromatin accessibility complex 1|Chromatin accessibility complex 15 kDa protein|Chromatin accessibility complex protein 1|DNA polymerase epsilon subunit p15|histone-fold protein CHRAC15|HuCHRAC15|YCL1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:100 to 1:300, ELISA:1:5000
Type:
Antigen: CHRAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse