Anti-BBOX1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: BBOX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-597
- 77819-596
- Antigen Symbol:
- BBOX1
- BBOX1
- Antigen Name:
- Gamma-butyrobetaine dioxygenase protein (154-247AA)
- Gamma-butyrobetaine dioxygenase protein (154-247AA)
- Antigen Synonyms:
- 2-oxoglutarate dioxygenase|BBH|BBOX 1|BBOX|Bbox1|BODG_HUMAN|Butyrobetaine (gamma) 2 oxoglutarate dioxygenase (gamma butyrobetaine hydroxylase) 1|G BBH|Gamma BBH|Gamma butyrobetaine 2 oxoglutarate dioxygenase 1|Gamma butyrobetaine dioxygenase|Gamma butyrobetaine hydroxylase|Gamma-BBH|Gamma-butyrobetaine|Gamma-butyrobetaine dioxygenase|Gamma-butyrobetaine hydroxylase
- 2-oxoglutarate dioxygenase|BBH|BBOX 1|BBOX|Bbox1|BODG_HUMAN|Butyrobetaine (gamma) 2 oxoglutarate dioxygenase (gamma butyrobetaine hydroxylase) 1|G BBH|Gamma BBH|Gamma butyrobetaine 2 oxoglutarate dioxygenase 1|Gamma butyrobetaine dioxygenase|Gamma butyrobetaine hydroxylase|Gamma-BBH|Gamma-butyrobetaine|Gamma-butyrobetaine dioxygenase|Gamma-butyrobetaine hydroxylase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75936
- O75936
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together