Order Entry
United States
ContactUsLinkComponent
Anti-BCL11B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-BCL11B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-BCL11B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77840-466
  • 77840-467
  • Antigen Symbol:
  • BCL11B
  • BCL11B
  • Antigen Name:
  • B-cell lymphoma/leukemia 11B protein (500-550AA)
  • B-cell lymphoma/leukemia 11B protein (500-550AA)
  • Antigen Synonyms:
  • ATL1 alpha|ATL1|ATL1 beta|ATL1 gamma|ATL1-delta|B cell CLL/lymphoma 11B/T cell receptor delta constant region fusion protein|B cell lymphoma/leukemia 11B|B-cell CLL/lymphoma 11B (zinc finger protein)|B-cell CLL/lymphoma 11B|B-cell lymphoma/leukemia 11B|BC11B_HUMAN|BCL-11B|Bcl11b|BCL11B/TRDC fusion|COUP TF interacting protein 2|COUP-TF interacting protein 2|COUP-TF-interacting protein 2|Ctip 2|CTIP-2|CTIP2|hRIT1 alpha|hRit1|Radiation induced tumor suppressor gene 1|Radiation induced tumor suppressor gene 1 protein|Radiation-induced tumor suppressor gene 1 protein|Rit 1|Rit1|zinc finger protein hRit1 alpha|ZNF856B
  • ATL1 alpha|ATL1|ATL1 beta|ATL1 gamma|ATL1-delta|B cell CLL/lymphoma 11B/T cell receptor delta constant region fusion protein|B cell lymphoma/leukemia 11B|B-cell CLL/lymphoma 11B (zinc finger protein)|B-cell CLL/lymphoma 11B|B-cell lymphoma/leukemia 11B|BC11B_HUMAN|BCL-11B|Bcl11b|BCL11B/TRDC fusion|COUP TF interacting protein 2|COUP-TF interacting protein 2|COUP-TF-interacting protein 2|Ctip 2|CTIP-2|CTIP2|hRIT1 alpha|hRit1|Radiation induced tumor suppressor gene 1|Radiation induced tumor suppressor gene 1 protein|Radiation-induced tumor suppressor gene 1 protein|Rit 1|Rit1|zinc finger protein hRit1 alpha|ZNF856B
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9C0K0
  • Q9C0K0
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together