Anti-SEPT9 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: SEPT9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Specifications
- Catalog No:
- 77813-948
- 77813-949
- Antigen Symbol:
- SEPT9
- SEPT9
- Antigen Name:
- Fusion protein of SEPT9
- Fusion protein of SEPT9
- Antigen Synonyms:
- AF17q25|Cell division control protein septin D1|KIAA0991|MLL septin like fusion protein|MLL septin-like fusion protein|MLL septin-like fusion protein MSF-A|MSF|MSF1|NAPB|Ov/Br septin|Ovarian/breast septin alpha|Ovarian/Breast septin|PNUTL4|SEPT9|SEPT9_HUMAN|SeptD1|Septin 9|Septin D1|Septin-9|SINT1
- AF17q25|Cell division control protein septin D1|KIAA0991|MLL septin like fusion protein|MLL septin-like fusion protein|MLL septin-like fusion protein MSF-A|MSF|MSF1|NAPB|Ov/Br septin|Ovarian/breast septin alpha|Ovarian/Breast septin|PNUTL4|SEPT9|SEPT9_HUMAN|SeptD1|Septin 9|Septin D1|Septin-9|SINT1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHD8
- Q9UHD8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together