Anti-CLEC6A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:2000, IHC:1:25 to 1:100
Type:
Antigen: CLEC6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77820-909
- 77820-908
- Antigen Symbol:
- CLEC6A
- CLEC6A
- Antigen Name:
- Synthetic peptide of CLEC6A
- Synthetic peptide of CLEC6A
- Antigen Synonyms:
- C type lectin domain family 6 member A|C type lectin superfamily member 10|C-type (calcium dependent|carbohydrate-recognition domain) lectin|superfamily member 10|C-type lectin domain family 6 member A|C-type lectin superfamily member 10|CLC6A_HUMAN|CLEC4N|mouse|homolog of|Clec6a|CLECSF10|DC associated C type lectin 2|DC-associated C-type lectin 2|Dectin-2|Dectin2|Dendritic cell associated lectin 2|Dendritic cell-associated C-type lectin 2
- C type lectin domain family 6 member A|C type lectin superfamily member 10|C-type (calcium dependent|carbohydrate-recognition domain) lectin|superfamily member 10|C-type lectin domain family 6 member A|C-type lectin superfamily member 10|CLC6A_HUMAN|CLEC4N|mouse|homolog of|Clec6a|CLECSF10|DC associated C type lectin 2|DC-associated C-type lectin 2|Dectin-2|Dectin2|Dendritic cell associated lectin 2|Dendritic cell-associated C-type lectin 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6EIG7
- Q6EIG7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together