Anti-RNF139 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: RNF139
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77813-642
- 77813-643
- Antigen Symbol:
- RNF139
- RNF139
- Antigen Name:
- Synthetic peptide of RNF139
- Synthetic peptide of RNF139
- Antigen Synonyms:
- TRC8|E3 ubiquitin-protein ligase RNF139|EC 2.3.2.27|RING finger protein 139|RING-type E3 ubiquitin transferase RNF139|Translocation in renal carcinoma on chromosome 8 protein
- TRC8|E3 ubiquitin-protein ligase RNF139|EC 2.3.2.27|RING finger protein 139|RING-type E3 ubiquitin transferase RNF139|Translocation in renal carcinoma on chromosome 8 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WU17
- Q8WU17
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together