Anti-CALD1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:1000 to 1:5000, IHC: 1:50 to 1:150
Type:
Antigen: CALD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77818-335
- 77818-334
- Antigen Symbol:
- CALD1
- CALD1
- Antigen Name:
- Fusion protein of CALD1
- Fusion protein of CALD1
- Antigen Synonyms:
- CAD| CALD 1| CALD1| CALD1_HUMAN| Caldesmon 1| Caldesmon 1 Isoform 1| Caldesmon 1 Isoform 2| Caldesmon 1 Isoform 3| Caldesmon 1 Isoform 4| Caldesmon 1 Isoform 5| Caldesmon| Caldesmon1| CDM| H CAD| HCAD| L CAD| LCAD| MGC21352| NAG22
- CAD| CALD 1| CALD1| CALD1_HUMAN| Caldesmon 1| Caldesmon 1 Isoform 1| Caldesmon 1 Isoform 2| Caldesmon 1 Isoform 3| Caldesmon 1 Isoform 4| Caldesmon 1 Isoform 5| Caldesmon| Caldesmon1| CDM| H CAD| HCAD| L CAD| LCAD| MGC21352| NAG22
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q05682
- Q05682
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together