Anti-NDRG2 Rabbit Polyclonal Antibody
Catalog # 77807-230
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of NDRG2.
- Antigen Symbol:NDRG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UN36
- Antigen Synonyms:Antidepressant related protein ADRG123|Cytoplasmic protein Ndr1|DKFZp781G1938|FLJ25522|KIAA1248|N myc downstream regulated gene 2|N myc downstream regulator 2|NDR1 related protein NDR2|NDRG 2|NDRG family member 2|NDRG1 related protein|NDRG2|NDRG2_HUMAN|Protein NDRG2|Protein Syld709613|SYLD|Syld709613 protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: NDRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat