Anti-NDUFB1 Rabbit Polyclonal Antibody
Catalog # 77797-646
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the N-terminal region of NDUFB1.
- Antigen Symbol:NDUFB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75438
- Antigen Synonyms:CI-MNLL|CI-SGDH|complex I MNLL subunit|Complex I-MNLL|MNLL|NADH dehydrogenase (ubiquinone) 1 beta subcomplex|1|7kDa|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1|NADH-ubiquinone oxidoreductase 1 beta subcomplex|NADH-ubiquinone oxidoreductase MNLL subunit|NADH:ubiquinone oxidoreductase subunit B1|NDUB1_HUMAN|NDUFB1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: NDUFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human