Anti-PITPNM1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PITPNM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-609
- 77797-608
- Antigen Symbol:
- PITPNM1
- PITPNM1
- Antigen Name:
- Membrane-associated phosphatidylinositol transfer protein 1 protein (1-238AA)
- Membrane-associated phosphatidylinositol transfer protein 1 protein (1-238AA)
- Antigen Synonyms:
- DRES9|Drosophila retinal degeneration B|Drosophila retinal degeneration B homolog|FLJ44997|membrane-associated 1|Membrane-associated phosphatidylinositol transfer protein 1|NIR 2|NIR-2|NIR2|OTTHUMP00000236414|OTTHUMP00000236415|Phosphatidylinositol transfer protein|phosphatidylinositol transfer protein membrane associated|PITM1_HUMAN|PITPnm 1|PITPNM|Pitpnm1|PYK2 N terminal domain interacting receptor 2|Pyk2 N-terminal domain-interacting receptor 2|RD 9|RD9|RDGB|RDGB1|RDGBA1|retinal degeneration B alpha 1
- DRES9|Drosophila retinal degeneration B|Drosophila retinal degeneration B homolog|FLJ44997|membrane-associated 1|Membrane-associated phosphatidylinositol transfer protein 1|NIR 2|NIR-2|NIR2|OTTHUMP00000236414|OTTHUMP00000236415|Phosphatidylinositol transfer protein|phosphatidylinositol transfer protein membrane associated|PITM1_HUMAN|PITPnm 1|PITPNM|Pitpnm1|PYK2 N terminal domain interacting receptor 2|Pyk2 N-terminal domain-interacting receptor 2|RD 9|RD9|RDGB|RDGB1|RDGBA1|retinal degeneration B alpha 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00562
- O00562
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together