Anti-JMJD1C Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: JMJD1C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-485
- 77825-486
- Antigen Symbol:
- JMJD1C
- JMJD1C
- Antigen Name:
- Probable JmjC domain-containing histone demethylation protein 2C protein (539-648AA)
- Probable JmjC domain-containing histone demethylation protein 2C protein (539-648AA)
- Antigen Synonyms:
- JHD2C_HUMAN|Jmjd1c|Jumonji domain containing 1C |Jumonji domain-containing protein 1C|Probable JmjC domain-containing histone demethylation protein 2C|Thryoid receptor interacting protein|Thyroid receptor-interacting protein 8|TR-interacting protein 8|TRIP-8|TRIP8
- JHD2C_HUMAN|Jmjd1c|Jumonji domain containing 1C |Jumonji domain-containing protein 1C|Probable JmjC domain-containing histone demethylation protein 2C|Thryoid receptor interacting protein|Thyroid receptor-interacting protein 8|TR-interacting protein 8|TRIP-8|TRIP8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15652
- Q15652
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together