Anti-DHX30 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DHX30
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-475
- 77830-476
- Antigen Symbol:
- DHX30
- DHX30
- Antigen Name:
- Putative ATP-dependent RNA helicase DHX30 protein (694-1000AA)
- Putative ATP-dependent RNA helicase DHX30 protein (694-1000AA)
- Antigen Synonyms:
- DDX30|DEAH box protein 30|Dhx30|_HUMAN|FLJ11214|KIAA0890 |Putative ATP-dependent RNA helicase |Ret CoR
- DDX30|DEAH box protein 30|Dhx30|_HUMAN|FLJ11214|KIAA0890 |Putative ATP-dependent RNA helicase |Ret CoR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L2E3
- Q7L2E3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together