Anti-SERPINH1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: SERPINH1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-223
- 77799-224
- Antigen Symbol:
- SERPINH1
- SERPINH1
- Antigen Name:
- Serpin H1 protein (139-418AA)
- Serpin H1 protein (139-418AA)
- Antigen Synonyms:
- 47 kDa heat shock protein|47 kDa heat shock protein precursor|Arsenic transactivated protein 3|Arsenic-transactivated protein 3|AsTP 3|AsTP3|CBP 1|CBP 2|CBP1|CBP2|Cell proliferation-inducing gene 14 protein|Collagen binding protein 1|Collagen binding protein 2|Collagen binding protein|Collagen-binding protein 2|Collagen-binding protein|Colligen|Colligin 1|Colligin 2|Colligin|colligin-1|colligin-2|gp46|Heat shock protein 47|Heat-shock protein 47|HGNC 1547|Hsp 47|J6|OI10|PIG 14|PIG14|PPROM|Proliferation inducing gene 14|Proliferation inducing gene 14 protein|RA A47|RA-A47|Rheumatoid arthritis antigen A 47|rheumatoid arthritis antigen A-47|Rheumatoid arthritis related antigen RA A47|Rheumatoid arthritis-related antigen RA-A47|serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)|serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2)|Serine or cysteine proteinase inhibitor clade H member 1|Serine or cysteine proteinase inhibitor clade H member 2|SERPH_HUMAN|Serpin H1|Serpin peptidase inhibitor clade H member 1|serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)|Serpin peptidase inhibitor, clade H, member 1|SERPINH1|SERPINH2
- 47 kDa heat shock protein|47 kDa heat shock protein precursor|Arsenic transactivated protein 3|Arsenic-transactivated protein 3|AsTP 3|AsTP3|CBP 1|CBP 2|CBP1|CBP2|Cell proliferation-inducing gene 14 protein|Collagen binding protein 1|Collagen binding protein 2|Collagen binding protein|Collagen-binding protein 2|Collagen-binding protein|Colligen|Colligin 1|Colligin 2|Colligin|colligin-1|colligin-2|gp46|Heat shock protein 47|Heat-shock protein 47|HGNC 1547|Hsp 47|J6|OI10|PIG 14|PIG14|PPROM|Proliferation inducing gene 14|Proliferation inducing gene 14 protein|RA A47|RA-A47|Rheumatoid arthritis antigen A 47|rheumatoid arthritis antigen A-47|Rheumatoid arthritis related antigen RA A47|Rheumatoid arthritis-related antigen RA-A47|serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)|serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2)|Serine or cysteine proteinase inhibitor clade H member 1|Serine or cysteine proteinase inhibitor clade H member 2|SERPH_HUMAN|Serpin H1|Serpin peptidase inhibitor clade H member 1|serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)|Serpin peptidase inhibitor, clade H, member 1|SERPINH1|SERPINH2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50454
- P50454
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together