Anti-REG3G Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: REG3G
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-358
- 77830-359
- Antigen Symbol:
- REG3G
- REG3G
- Antigen Name:
- Regenerating islet-derived protein 3-gamma protein (27-175AA)
- Regenerating islet-derived protein 3-gamma protein (27-175AA)
- Antigen Synonyms:
- LPPM429|Pancreatitis-associated protein 1B|Pancreatitis-associated protein IB|PAP IB|PAP-1B|PAP1B|PAPIB|REG III|Reg III-gamma|REG-3-gamma||_HUMAN|Regenerating islet derived 3 gamma|Regenerating islet-derived protein 3-gamma|Regenerating islet-derived protein III-gamma|UNQ429
- LPPM429|Pancreatitis-associated protein 1B|Pancreatitis-associated protein IB|PAP IB|PAP-1B|PAP1B|PAPIB|REG III|Reg III-gamma|REG-3-gamma||_HUMAN|Regenerating islet derived 3 gamma|Regenerating islet-derived protein 3-gamma|Regenerating islet-derived protein III-gamma|UNQ429
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UW15
- Q6UW15
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together