Anti-DDX53 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: DDX53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77816-439
- 77816-440
- Antigen Symbol:
- DDX53
- DDX53
- Antigen Name:
- Fusion protein of DDX53
- Fusion protein of DDX53
- Antigen Synonyms:
- CAGE|Cancer associated gene protein|Cancer-associated gene protein|Cancer/testis antigen 26|CT26|DDX53_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 53|DEAD box protein 53|DEAD box protein CAGE|Probable ATP dependent RNA helicase Probable ATP-dependent RNA helicase DDX53
- CAGE|Cancer associated gene protein|Cancer-associated gene protein|Cancer/testis antigen 26|CT26|DDX53_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 53|DEAD box protein 53|DEAD box protein CAGE|Probable ATP dependent RNA helicase Probable ATP-dependent RNA helicase DDX53
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86TM3
- Q86TM3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together