Anti-MACC1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MACC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-254
- 77831-253
- Antigen Symbol:
- MACC1
- MACC1
- Antigen Name:
- Metastasis-associated in colon cancer protein 1 protein (371-514AA)
- Metastasis-associated in colon cancer protein 1 protein (371-514AA)
- Antigen Synonyms:
- 7A5|MACC1|MACC1_HUMAN|Metastasis associated in colon cancer 1|Metastasis-associated in colon cancer protein 1|OTTHUMP00000201891|Putative binding protein 7a5|SH3 domain-containing protein 7a5|SH3BP4L
- 7A5|MACC1|MACC1_HUMAN|Metastasis associated in colon cancer 1|Metastasis-associated in colon cancer protein 1|OTTHUMP00000201891|Putative binding protein 7a5|SH3 domain-containing protein 7a5|SH3BP4L
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZN28
- Q6ZN28
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together