Anti-THAP12 Rabbit Recombinant Polyclonal Antibody
Catalog # 77795-404
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:52 kDa repressor of the inhibitor of the protein kinase protein (612-761AA)
- Antigen Symbol:THAP12
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O43422
- Antigen Synonyms:THAP12|DAP4|P52RIPK|PRKRIR|THAP0|52 kDa repressor of the inhibitor of the protein kinase|p52rIPK|58 kDa interferon-induced protein kinase-interacting protein|p58IPK-interacting protein|Death-associated protein 4|THAP domain-containing protein 0|THAP domain-containing protein 12
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: THAP12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human