Anti-RPS6KA5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RPS6KA5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-209
- 77798-208
- Antigen Symbol:
- RPS6KA5
- RPS6KA5
- Antigen Name:
- Ribosomal protein S6 kinase alpha-5 protein (224-367AA)
- Ribosomal protein S6 kinase alpha-5 protein (224-367AA)
- Antigen Synonyms:
- 90 kDa ribosomal protein S6 kinase 5|EC 2.7.11.1|KS6A5_HUMAN|MGC1911|Mitogen and stress activated protein kinase 1|MSPK1|Nuclear Mitogen And Stress Activated Protein Kinase 1|Nuclear mitogen- and stress-activated protein kinase 1|Ribosomal protein S6 kinase 90kD polypeptide 5|Ribosomal protein S6 kinase 90kDa|Ribosomal protein S6 kinase 90kDa polypeptide 5|Ribosomal Protein S6 Kinase Alpha 5|Ribosomal protein S6 kinase alpha-5|RLPK|RPS6KA5|RSK Like Protein Kinase|RSK-like protein kinase|RSKL|S6K alpha 5|S6K-alpha-5
- 90 kDa ribosomal protein S6 kinase 5|EC 2.7.11.1|KS6A5_HUMAN|MGC1911|Mitogen and stress activated protein kinase 1|MSPK1|Nuclear Mitogen And Stress Activated Protein Kinase 1|Nuclear mitogen- and stress-activated protein kinase 1|Ribosomal protein S6 kinase 90kD polypeptide 5|Ribosomal protein S6 kinase 90kDa|Ribosomal protein S6 kinase 90kDa polypeptide 5|Ribosomal Protein S6 Kinase Alpha 5|Ribosomal protein S6 kinase alpha-5|RLPK|RPS6KA5|RSK Like Protein Kinase|RSK-like protein kinase|RSKL|S6K alpha 5|S6K-alpha-5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75582
- O75582
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together