Anti-LTBP4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: LTBP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77798-258
- 77798-257
- Antigen Symbol:
- LTBP4
- LTBP4
- Antigen Name:
- Latent-transforming growth factor beta-binding protein 4 protein (977-1180AA)
- Latent-transforming growth factor beta-binding protein 4 protein (977-1180AA)
- Antigen Synonyms:
- latent transforming growth factor beta binding protein 4|latent transforming growth factor beta binding protein 4L|Latent-transforming growth factor beta-binding protein 4|LTBP 4|LTBP-4|LTBP4|LTBP4_HUMAN|LTBP4L|LTBP4S
- latent transforming growth factor beta binding protein 4|latent transforming growth factor beta binding protein 4L|Latent-transforming growth factor beta-binding protein 4|LTBP 4|LTBP-4|LTBP4|LTBP4_HUMAN|LTBP4L|LTBP4S
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N2S1
- Q8N2S1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together