Anti-CYP2D6 Rabbit Polyclonal Antibody
Catalog # 77806-026
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of Cytochrome P450 2D6.
- Antigen Symbol:CYP2D6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P10635
- Antigen Synonyms:Cholesterol 25-hydroxylase|CP2D6_HUMAN|CPD6|CYP2D|CYP2D6|CYP2D7AP|CYP2D7BP|CYP2D7P2|CYP2D8P2|CYP2DL1|CYPIID6|Cytochrome P450 2D6|Cytochrome P450 DB1|Cytochrome P450 family 2 subfamily D member 6|Cytochrome P450 family 2 subfamily D polypeptide 6|Cytochrome P450-DB1|Debrisoquine 4 hydroxylase|Debrisoquine 4-hydroxylase|Flavoprotein linked monooxygenase|Microsomal monooxygenase|P450 DB1|P450C2D|P450DB1|Xenobiotic monooxygenase
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100
Type:
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human