Anti-ARFRP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARFRP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-355
- 77824-354
- Antigen Symbol:
- ARFRP1
- ARFRP1
- Antigen Name:
- ADP-ribosylation factor-related protein 1 protein (1-201AA)
- ADP-ribosylation factor-related protein 1 protein (1-201AA)
- Antigen Synonyms:
- ADP ribosylation factor related protein 1|ADP-ribosylation factor-related protein 1|ARF related protein 1|ARF related protein|ARF-related protein 1|ARFRP_HUMAN|ARFRP1|ARL 18|ARL18|ARP 1|ARP|ARP1|Helicase like protein NHL|MGC6837|SCG10 like protein
- ADP ribosylation factor related protein 1|ADP-ribosylation factor-related protein 1|ARF related protein 1|ARF related protein|ARF-related protein 1|ARFRP_HUMAN|ARFRP1|ARL 18|ARL18|ARP 1|ARP|ARP1|Helicase like protein NHL|MGC6837|SCG10 like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13795
- Q13795
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together