Anti-TRIM8 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: TRIM8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77838-176
- 77838-175
- Antigen Symbol:
- TRIM8
- TRIM8
- Antigen Name:
- Probable E3 ubiquitin-protein ligase TRIM8 protein (367-517AA)
- Probable E3 ubiquitin-protein ligase TRIM8 protein (367-517AA)
- Antigen Synonyms:
- AA408830| BB006755| GERP| Glioblastoma-expressed RING finger protein| Lioblastoma expressed ring finger protein| Probable E3 ubiquitin-protein ligase | RING finger protein 27| RNF27| RP11-47A8.4| | _HUMAN| Tripartite motif containing 8| Tripartite motif protein | Tripartite motif-containing protein 8
- AA408830| BB006755| GERP| Glioblastoma-expressed RING finger protein| Lioblastoma expressed ring finger protein| Probable E3 ubiquitin-protein ligase | RING finger protein 27| RNF27| RP11-47A8.4| | _HUMAN| Tripartite motif containing 8| Tripartite motif protein | Tripartite motif-containing protein 8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZR9
- Q9BZR9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together