Anti-MED17 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MED17
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-556
- 77836-555
- Antigen Symbol:
- MED17
- MED17
- Antigen Name:
- Mediator of RNA polymerase II transcription subunit 17 protein (411-651AA)
- Mediator of RNA polymerase II transcription subunit 17 protein (411-651AA)
- Antigen Synonyms:
- 77-KD subunit|Activator recruited cofactor 77 kDa component|Activator-recruited cofactor 77 kDa component|ARC77|Cofactor required for Sp1 transcriptional activation subunit 6|cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa|CRSP complex subunit 6|CRSP6|CRSP77|DRIP80|med17|MED17_HUMAN|Mediator complex subunit 17|Mediator of RNA polymerase II transcription subunit 17|Mediator of RNA polymerase II transcription subunit 17, S cerevisiae, homolog of|Thyroid hormone receptor-associated protein complex 80 kDa component|Thyroid hormone receptor-associated protein, 80-KD subunit|Transcriptional coactivator CRSP77|Trap80|VITAMIN D RECEPTOR-INTERACTING PROTEIN, 80-KD|Vitamin D3 receptor-interacting protein complex 80 kDa component
- 77-KD subunit|Activator recruited cofactor 77 kDa component|Activator-recruited cofactor 77 kDa component|ARC77|Cofactor required for Sp1 transcriptional activation subunit 6|cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa|CRSP complex subunit 6|CRSP6|CRSP77|DRIP80|med17|MED17_HUMAN|Mediator complex subunit 17|Mediator of RNA polymerase II transcription subunit 17|Mediator of RNA polymerase II transcription subunit 17, S cerevisiae, homolog of|Thyroid hormone receptor-associated protein complex 80 kDa component|Thyroid hormone receptor-associated protein, 80-KD subunit|Transcriptional coactivator CRSP77|Trap80|VITAMIN D RECEPTOR-INTERACTING PROTEIN, 80-KD|Vitamin D3 receptor-interacting protein complex 80 kDa component
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVC6
- Q9NVC6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together