About this item
Recommended dilution: ELISA:1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: TBL1X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77818-479
- 77818-480
- Antigen Symbol:
- TBL1X
- TBL1X
- Antigen Name:
- Fusion protein of TBL1X
- Fusion protein of TBL1X
- Antigen Synonyms:
- EBI|F box like/WD repeat protein TBL1X|F-box-like/WD repeat-containing protein TBL1X|SMAP 55|SMAP55|TBL 1|TBL 1X|TBL1|TBL1X|TBL1X_HUMAN|Transducin (beta) like 1|Transducin (beta) like 1 X linked|Transducin (beta) like 1X linked|Transducin beta like 1 X linked|Transducin beta like 1X|Transducin beta like 1X protein|Transducin beta like protein 1, X linked|Transducin beta-like protein 1X|Transducin-beta-like protein 1|X-linked
- EBI|F box like/WD repeat protein TBL1X|F-box-like/WD repeat-containing protein TBL1X|SMAP 55|SMAP55|TBL 1|TBL 1X|TBL1|TBL1X|TBL1X_HUMAN|Transducin (beta) like 1|Transducin (beta) like 1 X linked|Transducin (beta) like 1X linked|Transducin beta like 1 X linked|Transducin beta like 1X|Transducin beta like 1X protein|Transducin beta like protein 1, X linked|Transducin beta-like protein 1X|Transducin-beta-like protein 1|X-linked
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60907
- O60907
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice