Anti-MAGI2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAGI2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-183
- 77832-182
- Antigen Symbol:
- MAGI2
- MAGI2
- Antigen Name:
- Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2 protein (1308-1455AA)
- Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2 protein (1308-1455AA)
- Antigen Synonyms:
- Activin receptor interacting protein 1|Acvri1 |Acvrinp1|ACVRIP1|AIP 1|Aip-1|ARIP1|Atrophin 1 interacting protein 1|Atrophin 1 interacting protein A|Atrophin-1-interacting protein 1|Atrophin-1-interacting protein A|KIAA0705|MAGI-2||_HUMAN|Membrane associated guanylate kinase 2|Membrane associated guanylate kinase inverted 2|Membrane associated guanylate kinase WW and PDZ domain containing 2|Membrane associated guanylate kinase WW and PDZ domain containing protein 2|Membrane-associated guanylate kinase|Membrane-associated guanylate kinase inverted 2|S SCAM |SSCAM|WW and PDZ domain-containing protein 2
- Activin receptor interacting protein 1|Acvri1 |Acvrinp1|ACVRIP1|AIP 1|Aip-1|ARIP1|Atrophin 1 interacting protein 1|Atrophin 1 interacting protein A|Atrophin-1-interacting protein 1|Atrophin-1-interacting protein A|KIAA0705|MAGI-2||_HUMAN|Membrane associated guanylate kinase 2|Membrane associated guanylate kinase inverted 2|Membrane associated guanylate kinase WW and PDZ domain containing 2|Membrane associated guanylate kinase WW and PDZ domain containing protein 2|Membrane-associated guanylate kinase|Membrane-associated guanylate kinase inverted 2|S SCAM |SSCAM|WW and PDZ domain-containing protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UL8
- Q86UL8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together