Anti-TSPAN8 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TSPAN8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-649
- 77811-649
- Antigen Symbol:
- TSPAN8
- TSPAN8
- Antigen Name:
- Tetraspanin-8 protein (110-205AA)
- Tetraspanin-8 protein (110-205AA)
- Antigen Synonyms:
- anti TSPAN8|CO029|Tetraspanin 8|Tetraspanin-8|TM4SF3|Transmembrane 4 superfamily member 3|TSN8_HUMAN|Tspan 8|Tspan-8|TSPAN8|Tumor associated antigen CO 029|Tumor-associated antigen CO-029
- anti TSPAN8|CO029|Tetraspanin 8|Tetraspanin-8|TM4SF3|Transmembrane 4 superfamily member 3|TSN8_HUMAN|Tspan 8|Tspan-8|TSPAN8|Tumor associated antigen CO 029|Tumor-associated antigen CO-029
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19075
- P19075
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together