Anti-XPR1 Rabbit Polyclonal Antibody
Catalog # 77804-759
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:XPR1_HUMAN
- Antigen Symbol:XPR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UBH6
- Antigen Synonyms:FLJ90308|OTTHUMP00000033084|OTTHUMP00000033085|Polytropic murine leukamia virus receptor SYG 1|Polytropic murine leukamia virus receptor SYG1|Protein SYG1 homolog|SYG 1|SYG1|SYG1 protein|X-receptor|X3|Xenotropic and polytropic murine leukemia virus receptor X3|Xenotropic and polytropic retrovirus receptor|Xenotropic and polytropic retrovirus receptor 1|XPR 1|XPR1|XPR1 protein|XPR1_HUMAN
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: XPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat