Anti-MKRN2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: MKRN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77838-236
- 77838-235
- Antigen Symbol:
- MKRN2
- MKRN2
- Antigen Name:
- Probable E3 ubiquitin-protein ligase makorin-2 protein (29-170AA)
- Probable E3 ubiquitin-protein ligase makorin-2 protein (29-170AA)
- Antigen Synonyms:
- RNF62|HSPC070Probable E3 ubiquitin-protein ligase makorin-2|EC 2.3.2.27|RING finger protein 62|RING-type E3 ubiquitin transferase makorin-2
- RNF62|HSPC070Probable E3 ubiquitin-protein ligase makorin-2|EC 2.3.2.27|RING finger protein 62|RING-type E3 ubiquitin transferase makorin-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H000
- Q9H000
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together