Anti-P2RY14 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IP: 1:200 to 1:2000
Type:
Antigen: P2RY14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77825-461
- 77825-462
- Antigen Symbol:
- P2RY14
- P2RY14
- Antigen Name:
- P2Y purinoceptor 14 protein (209-338AA)
- P2Y purinoceptor 14 protein (209-338AA)
- Antigen Synonyms:
- A330108O13Rik|G protein coupled receptor for UDP-glucose|G protein-coupled receptor 105|G protein-coupled receptor VTR 15-20|G-protein coupled receptor 105|GPCR GPR105|GPR105|KIAA0001|P2RY14|P2Y purinoceptor 14|P2Y(14) receptor|P2Y14|P2Y14 receptor|P2Y14_HUMAN|Purinergic receptor P2Y G protein coupled 14|Similar to TYPE-1 Angiotensin II receptor|UDP-glucose receptor
- A330108O13Rik|G protein coupled receptor for UDP-glucose|G protein-coupled receptor 105|G protein-coupled receptor VTR 15-20|G-protein coupled receptor 105|GPCR GPR105|GPR105|KIAA0001|P2RY14|P2Y purinoceptor 14|P2Y(14) receptor|P2Y14|P2Y14 receptor|P2Y14_HUMAN|Purinergic receptor P2Y G protein coupled 14|Similar to TYPE-1 Angiotensin II receptor|UDP-glucose receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15391
- Q15391
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together