Anti-SCO2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SCO2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-854
- 77798-855
- Antigen Symbol:
- SCO2
- SCO2
- Antigen Name:
- Protein SCO2 homolog, mitochondrial protein (1-130AA)
- Protein SCO2 homolog, mitochondrial protein (1-130AA)
- Antigen Synonyms:
- Cytochrome oxidase deficient homolog 2|MGC125823|MGC125825|OTTHUMP00000196774|OTTHUMP00000196775|Protein SCO2 homolog|mitochondrial|SCO (cytochrome oxidase deficient|yeast) homolog 2|SCO 1L|SCO 2|SCO cytochrome oxidase deficient homolog 2 (yeast)|SCO cytochrome oxidase deficient homolog 2|SCO1L|SCO2|SCO2_HUMAN|Synthesis of cytochrome c oxidase 2
- Cytochrome oxidase deficient homolog 2|MGC125823|MGC125825|OTTHUMP00000196774|OTTHUMP00000196775|Protein SCO2 homolog|mitochondrial|SCO (cytochrome oxidase deficient|yeast) homolog 2|SCO 1L|SCO 2|SCO cytochrome oxidase deficient homolog 2 (yeast)|SCO cytochrome oxidase deficient homolog 2|SCO1L|SCO2|SCO2_HUMAN|Synthesis of cytochrome c oxidase 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43819
- O43819
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together