Anti-CDC6 Rabbit Polyclonal Antibody
Catalog # 77812-466
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the C-terminal region of Cdc6.
- Antigen Symbol:CDC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q99741
- Antigen Synonyms:Cdc 18L|Cdc 6|CDC18 (cell division cycle 18|S.pombe|homolog) like|CDC18 (S.pombe)|Cdc18 related protein|CDC18(S.pombe)|Cdc18-related protein|Cdc18L|cdc6|CDC6 cell division cycle 6 homolog|CDC6 related protein|CDC6-related protein|CDC6_HUMAN|Cdc6p|CELL CYCLE CONTROLLER CDC6|Cell division control protein 6|Cell division control protein 6 homolog|Cell division cycle 6 homolog (S. cerevisiae)|Cell division cycle 6 homolog|Cell division cycle 6|S. cerevisiae|homolog of|HsCDC 6|HsCDC18|HsCDC6|p62|p62(cdc 6)|p62(cdc6)
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: CDC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human