Anti-RBPJ Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RBPJ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-877
- 77796-876
- Antigen Symbol:
- RBPJ
- RBPJ
- Antigen Name:
- Recombining binding protein suppressor of hairless protein (243-485AA)
- Recombining binding protein suppressor of hairless protein (243-485AA)
- Antigen Synonyms:
- AI843960|AOS3|CBF 1|CBF-1|CBF1|csl|IGKJRB|IGKJRB1|J kappa recombination signal binding protein|J kappa-recombination signal-binding protein|KBF2|NY REN 30 antigen|RBP J|RBP J kappa|RBP JK|RBP-J|RBP-J kappa|RBP-JK|Rbpj|RBPJK|RBPSUH|recombination signal binding protein for immunoglobulin kappa J region|Recombining binding protein suppressor of hairless|Renal carcinoma antigen NY-REN-30|SUH|SUH_HUMAN
- AI843960|AOS3|CBF 1|CBF-1|CBF1|csl|IGKJRB|IGKJRB1|J kappa recombination signal binding protein|J kappa-recombination signal-binding protein|KBF2|NY REN 30 antigen|RBP J|RBP J kappa|RBP JK|RBP-J|RBP-J kappa|RBP-JK|Rbpj|RBPJK|RBPSUH|recombination signal binding protein for immunoglobulin kappa J region|Recombining binding protein suppressor of hairless|Renal carcinoma antigen NY-REN-30|SUH|SUH_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q06330
- Q06330
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together