Anti-UBR5 Rabbit Polyclonal Antibody
Catalog # 77812-812
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from Internal of EDD
- Antigen Symbol:UBR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O95071
- Antigen Synonyms:4432411E13Rik|AW549941|C77315|D030042K14|DD5|E3 identified by differential display|E3 ubiquitin protein ligase, HECT domain containing, 1|E3 ubiquitin-protein ligase|E3 ubiquitin-protein ligase UBR5|EDD 1|EDD|EDD1|FLJ11310|HECT domain-containing 1|hHYD|HYD|Hyperplastic discs protein homolog|Hyperplastic discs, Drosophila, homolog of|KIAA0896|MGC57263|mKIAA0896|Progestin induced protein|Progestin-induced protein|Rat100|Ubiquitin protein ligase|Ubiquitin protein ligase E3 component n recognin 5|UBR5|UBR5_HUMAN
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100
Type:
Antigen: UBR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse