Anti-AP2S1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: AP2S1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77804-533
- 77804-532
- Antigen Symbol:
- AP2S1
- AP2S1
- Antigen Name:
- AP-2 complex subunit sigma protein (1-142AA)
- AP-2 complex subunit sigma protein (1-142AA)
- Antigen Synonyms:
- Adapter-related protein complex 2 sigma subunit|Adaptor protein complex AP 2 subunit sigma|Adaptor protein complex AP-2 subunit sigma|Adaptor protein complex AP2 subunit sigma|Adaptor related protein complex 2 sigma 1 subunit|AI043088|AP 17|AP 2 complex subunit sigma 1|AP-2 complex subunit sigma|AP17|AP17 delta|AP2 complex subunit sigma 1|Ap2s1|AP2S1_HUMAN|CLAPS 2|CLAPS2|Clathrin adaptor complex AP2|sigma subunit|Clathrin adaptor protein AP17|Clathrin assembly protein 2 small chain|Clathrin associated/assembly/adaptor protein small 2|17-KD|Clathrin associated/assembly/adaptor protein|small 2 (17kD)|Clathrin coat assembly protein AP17|Clathrin coat associated protein AP17|Clathrin coat-associated protein AP17|HA2 17 kDa subunit|MGC62945|Plasma membrane adaptor AP 2 17 kDa protein|Plasma membrane adaptor AP-2 17 kDa protein|Plasma membrane adaptor AP2 17 kDa protein|Sigma adaptin 3b|Sigma2 adaptin|Sigma2-adaptin
- Adapter-related protein complex 2 sigma subunit|Adaptor protein complex AP 2 subunit sigma|Adaptor protein complex AP-2 subunit sigma|Adaptor protein complex AP2 subunit sigma|Adaptor related protein complex 2 sigma 1 subunit|AI043088|AP 17|AP 2 complex subunit sigma 1|AP-2 complex subunit sigma|AP17|AP17 delta|AP2 complex subunit sigma 1|Ap2s1|AP2S1_HUMAN|CLAPS 2|CLAPS2|Clathrin adaptor complex AP2|sigma subunit|Clathrin adaptor protein AP17|Clathrin assembly protein 2 small chain|Clathrin associated/assembly/adaptor protein small 2|17-KD|Clathrin associated/assembly/adaptor protein|small 2 (17kD)|Clathrin coat assembly protein AP17|Clathrin coat associated protein AP17|Clathrin coat-associated protein AP17|HA2 17 kDa subunit|MGC62945|Plasma membrane adaptor AP 2 17 kDa protein|Plasma membrane adaptor AP-2 17 kDa protein|Plasma membrane adaptor AP2 17 kDa protein|Sigma adaptin 3b|Sigma2 adaptin|Sigma2-adaptin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53680
- P53680
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together