Order Entry
United States
ContactUsLinkComponent
Anti-CPT1C Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-CPT1C Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CPT1C Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77833-552
  • 77833-551
  • Antigen Symbol:
  • CPT1C
  • CPT1C
  • Antigen Name:
  • Carnitine O-palmitoyltransferase 1, brain isoform protein (699-803AA)
  • Carnitine O-palmitoyltransferase 1, brain isoform protein (699-803AA)
  • Antigen Synonyms:
  • brain isoform|Carnitine acyltransferase like protein 1|Carnitine acyltransferase like protein1|Carnitine O palmitoyltransferase 1 brain isoform|Carnitine O palmitoyltransferase I|Carnitine O-palmitoyltransferase 1|Carnitine O-palmitoyltransferase I|Carnitine palmitoyltransferase 1|brain|Carnitine palmitoyltransferase 1C|Carnitine palmitoyltransferase I related C|Carnitine palmitoyltransferase1C|CAT L1|CATL 1|CATL1|CPT 1 like pseudogene|Cpt 1c|CPT 1P|CPT I C|CPT IC|Cpt1 c|CPT1 like pseudogene|CPT1-B|Cpt1c|CPT1C_HUMAN|CPT1P|CPTI-B|CPTIC
  • brain isoform|Carnitine acyltransferase like protein 1|Carnitine acyltransferase like protein1|Carnitine O palmitoyltransferase 1 brain isoform|Carnitine O palmitoyltransferase I|Carnitine O-palmitoyltransferase 1|Carnitine O-palmitoyltransferase I|Carnitine palmitoyltransferase 1|brain|Carnitine palmitoyltransferase 1C|Carnitine palmitoyltransferase I related C|Carnitine palmitoyltransferase1C|CAT L1|CATL 1|CATL1|CPT 1 like pseudogene|Cpt 1c|CPT 1P|CPT I C|CPT IC|Cpt1 c|CPT1 like pseudogene|CPT1-B|Cpt1c|CPT1C_HUMAN|CPT1P|CPTI-B|CPTIC
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8TCG5
  • Q8TCG5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, protein G purified
  • >95%, protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together