To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:Synthesized peptide derived from MEK-1 around the phosphorylation site of T292.
- Antigen Symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q02750
- Antigen Synonyms:Dual specificity mitogen activated protein kinase kinase 1| Dual specificity mitogen-activated protein kinase kinase 1| ERK activator kinase 1| MAP kinase kinase 1| MAP kinase/Erk kinase 1| MAP2K1| MAPK/ERK kinase 1| MAPKK 1| MAPKK1| MEK 1| Mek1| MEKK1| Mitogen activated protein kinase kinase 1| MKK 1| MKK1| MP2K1_HUMAN| PRKMK1| Protein kinase mitogen activated kinase 1 (MAP kinase kinase 1)| Protein kinase mitogen activated, kinase 1| protein kinase mitogen-activated kinase 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:10000
Type:
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat