Anti-KRTAP3-3 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: KRTAP3-3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-232
- 77839-231
- Antigen Symbol:
- KRTAP3-3
- KRTAP3-3
- Antigen Name:
- Keratin-associated protein 3-3 protein (1-98AA)
- Keratin-associated protein 3-3 protein (1-98AA)
- Antigen Synonyms:
- KRTAP3-3|KAP3.3|KRTAP3.3Keratin-associated protein 3-3|High sulfur keratin-associated protein 3.3|Keratin-associated protein 3.3
- KRTAP3-3|KAP3.3|KRTAP3.3Keratin-associated protein 3-3|High sulfur keratin-associated protein 3.3|Keratin-associated protein 3.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BYR6
- Q9BYR6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together