Anti-TAP1 Rabbit Polyclonal Antibody
77801-848
New Product
- Antigen Name:Y3
- Antigen Symbol:TAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q03518
- Antigen Synonyms:ABC 17|ABC transporter MHC 1|ABC17|ABCB 2|ABCB2|Antigen peptide transporter 1|APT 1|APT1|ATP binding cassette sub family B (MDR/TAP) member 2|ATP binding cassette sub family B member 2|ATP binding cassette transporter|ATP-binding cassette sub-family B member 2|D6S114E|FLJ26666|FLJ41500|Peptide supply factor 1|Peptide transporter involved in antigen processing 1|Peptide transporter PSF 1|Peptide transporter PSF1|Peptide transporter TAP 1|Peptide transporter TAP1|PSF 1|PSF-1|PSF1|Really interesting new gene 4 protein|RING 4|RING4|TAP 1|TAP1|TAP1*0102N|TAP1_HUMAN|TAP1N|Transporter 1 ATP binding cassette sub family B (MDR/TAP)|Transporter 1 ATP binding cassette sub family B|Transporter associated with antigen processing|Transporter ATP binding cassette major histocompatibility complex 1|Y3
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: TAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat