Anti-SLAMF6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLAMF6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-687
- 77799-686
- Antigen Symbol:
- SLAMF6
- SLAMF6
- Antigen Name:
- SLAM family member 6 protein (248-331AA)
- SLAM family member 6 protein (248-331AA)
- Antigen Synonyms:
- Activating NK receptor|CD352|KALI|KALIb|LY108|MGC104953|Natural killer T and B cell antigen|NK T B antigen|NK-|T-|and B-cell antigen|NK-T-B-antigen|NTB A|NTB-A|NTBA|NTBA receptor|SF2000|SLAF6_HUMAN|SLAM family member 6|SLAM family member 6 precursor|SLAM Family Receptor|Slamf6
- Activating NK receptor|CD352|KALI|KALIb|LY108|MGC104953|Natural killer T and B cell antigen|NK T B antigen|NK-|T-|and B-cell antigen|NK-T-B-antigen|NTB A|NTB-A|NTBA|NTBA receptor|SF2000|SLAF6_HUMAN|SLAM family member 6|SLAM family member 6 precursor|SLAM Family Receptor|Slamf6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96DU3
- Q96DU3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together