Order Entry
United States
ContactUsLinkComponent
Anti-AZI2 Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-AZI2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-AZI2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77836-224
  • 77836-225
  • Antigen Symbol:
  • AZI2
  • AZI2
  • Antigen Name:
  • 5-azacytidine-induced protein 2 protein (258-376AA)
  • 5-azacytidine-induced protein 2 protein (258-376AA)
  • Antigen Synonyms:
  • 5 azacytidine induced protein 2|5-azacytidine-induced protein 2|AA410145|AZ2|AZI2|AZI2_HUMAN|MGC112644|Nak associated protein 1|Nak-associated protein 1|NAP1|NF kappa B activating kinase associated protein 1|NF-kappa-B-activating kinase-associated protein 1|TILP|TILP(S)
  • 5 azacytidine induced protein 2|5-azacytidine-induced protein 2|AA410145|AZ2|AZI2|AZI2_HUMAN|MGC112644|Nak associated protein 1|Nak-associated protein 1|NAP1|NF kappa B activating kinase associated protein 1|NF-kappa-B-activating kinase-associated protein 1|TILP|TILP(S)
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9H6S1
  • Q9H6S1
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together